missing translation for 'onlineSavingsMsg'
Learn More

PGRMC1 Mouse anti-Human, Mouse, Rat, Clone: 1C10D11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66372-1-IG-20UL

 View more versions of this product

Product Code. 16898185

  • 1696.32 NOK / 20µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis.
Specifications

Specifications

PGRMC1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00264, O55022, P70580
Pgrmc1
PGRMC1 Fusion Protein Ag3643
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
1C10D11
Unconjugated
PGRMC1
HPR6.6, MPR, PGRMC, PGRMC1
Mouse
Protein G
RUO
10857, 291948, 53328
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.