missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN5 Rabbit anti-Human, Mouse, Rat, Clone: 9Y5H7, Novus Biologicals™
Rabbit Monoclonal Antibody
1815.00 NOK - 4580.00 NOK
Specifications
Antigen | PFDN5 |
---|---|
Clone | 9Y5H7 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18354372
|
Bio-Techne
NBP3-16331-20UL |
20 μg |
1815.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18329754
|
Bio-Techne
NBP3-16331-100UL |
100 μg |
4580.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PFDN5 Monoclonal antibody specifically detects PFDN5 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PFDN5 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5204 | |
IgG | |
Affinity purified |
9Y5H7 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
c-myc binding protein, C-Myc-binding protein Mm-1, MM-1, MM1MGC71907, Myc modulator 1, myc modulator-1, PFD5MGC5329, prefoldin 5, prefoldin subunit 5 | |
A synthetic peptide corresponding to a sequence within amino acids 50-154 of human PFDN5 (Q99471). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title