Learn More
BrandTech™ VITLAB™ PFA Round Bottom Flasks
Take advantage of PFA, which is especially suitable for use in trace analysis, with Vitlab™ PFA Round Bottom Flasks. They have high thermal stability, from -200 °C to +260 °C, and are autoclavable.
1355.00 NOK - 1865.00 NOK
Specifications
Material | PFA |
---|---|
For Use With (Application) | Suitable for use in trace analysis |
No. per Pack | 1 |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10384311
|
BrandTech™
107997 |
500 mL |
1865.00 NOK
Each |
Estimated Shipment: 27-06-2025
Log in to see stock. |
||||||
10777991
|
BrandTech™
107897 |
250 mL |
1355.00 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
Description
- Transparent
- Neck with NS 29/32
- Suitable as safety flask
- For use with rotary evaporators (operation at room temperature) to collect distilled-off liquid.
- High thermal stability and chemical resistance
Specifications
PFA | |
Suitable for use in trace analysis | |
1 | |
Autoclavable |
Product Title
BrandTech™ VITLAB™ PFA Round Bottom Flasks