Learn More
VITLAB™ PFA Flask
Brand:  VITLAB™ 107797
Product Code. 10470043
  • 1160.00 NOK / Each
Estimated Shipment: 25-03-2025
to see stock.
Explore more special offers
Specifications
100 mL
PFA
Product Title
VITLAB™ PFA Flask > Capacity: 100mL