Learn More
Cole-Parmer™ PFA Compression Threaded Adapter
Excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
Brand:  Cole-Parmer™ 31321-45
Product Code. 11735089
  • 1070.00 NOK / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Superior chemical resistance with durable PFA composition.
  • Get a tight seal at higher pressures.
  • Withstand higher pressures than barbed fittings.
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications
Specifications
PFA compression threaded adapter, 10mm tubing OD x 3/8in male NPT
PFA
Autoclave or Ethylene Oxide
Compression x NPT(M)
10mm OD
3/8 in.