Description
- Sterile facemasks are individually packaged and lot traceable
- 10.0 to 20.0 kGy gamma irradiated
- Hypoallergenic, soft surface layers prevent skin irritation and allergy problems
- Silicon-free headloop OP bands
- Ultrasonic bonded loop bands ensure a cleaner mask, reducing contamination and outgassing
Specifications
Specifications
Product Type | Cleanroom Face Mask |
Color | White |
Material | Polypropylene, Rayon |
Sterility | Sterile |
Certifications/Compliance | Certificate of Irradiation; Certificate of Sterility; Certificate of Conformance; Lot traceable |
Construction | 3-ply |
Content And Storage | Five year, closed package expiration date. Individually packaged |
Disposable | Single-use |
Filter Efficiency | 94.74% per EN 14683:2019 |
For Use With (Application) | Cleanrooms |
Show More |
Product Content Correction
Product Title
Veltek PF-4 Sterile Cleanroom Facemask >