Learn More
PEX1 Mouse anti-Human, Unconjugated, Clone: 2F3H5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67436-1-IG-150UL
This item is not returnable.
View return policy
Description
The PEX1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PEX1. This antibody recognizes human, mouse, rat antigen. The PEX1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
PEX1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
2F3H5 | |
Unconjugated | |
150 μL | |
5189 | |
IgG2b |