Learn More
IKA™ PETTE Biosphere™ Plus M Pipette Tips
These Biosphere™ Plus M Pipette Tips feature a low retention filter tip and are sterile, free of DNA, DNase/RNase, PCR inhibitors, ATP, and pyrogens/endotoxins. Designed for use with IKA™ PETTE Pipettes.
Brand: IKA™ 0020100000
This item is not returnable.
View return policy
Description
- 1000 μL transparent pipette tips featuring calibration rings
- Low retention filter tip
- Composed of Biosphere™ Plus: sterile and free of DNA, DNase/RNase, PCR inhibitors, ATP, and pyrogens/endotoxins
- Designed for IKA™ PETTE Pipettes
Specifications
1000 μL | |
Sterile | |
Not Autoclavable | |
Box | |
1920 Tips (Total) | |
Single-use | |
PETTE Biosphere |
Filtered | |
Transparent | |
Calibration Rings | |
20 Trays (96 Tips per Tray) | |
Blue | |
IKA™ PETTE Pipettes | |
Biosphere™ Plus (sterile, free of DNA, DNase/RNase, PCR inhibitors, ATP, and pyrogens/endotoxins) |