missing translation for 'onlineSavingsMsg'
Learn More
Petroleum Ether, Boiling Range 40–60°C, ACS Reagent Grade, Scharlab Product Code.: 11368644
compliance-icons compliance-icons

Petroleum Ether, Boiling Range 40–60°C, ACS Reagent Grade, Scharlab

Product Code. 11368644
2.5 L
Click to view available options
Quantity:
2.5 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11368644

Brand: Scharlab ET00922500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Used in solvents in analytical chemistry.

  • Synonyms: Petroleum benzine, Petroleum spirit
  • CAS: 64742-49-0
  • EIN ECS No.: 265-151-9
  • Iodine Index: 0.3 max
  • Sulfur Compounds: 0.005% max
  • Aromatic Hydrocarbons (as C6H6): 0.02% max

Specifications

Melting Point <-100°C
CAS 64742-49-0
Acidity 0.0003mEq/g max
Appearance Clear
Density 0.65g/cm3 at 15°C
Boiling Point 40°C to 60°C (min 90% volume)
Quantity 2.5 L
Flash Point < -21°C
Ignition Point 250°C
Physical Form Liquid
Residue after Evaporation 0.0005% max
Molecular Formula C6H14
MDL Number MFCD00081849
Vapor Pressure 350hPa (at 20°C)
Water 0.01% max
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
Solubility Information Almost non-miscible (in water)
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
SMILES CCCC(C)C
IUPAC Name Petroleum ether
Molecular Weight (g/mol) 86.178
PubChem CID 7892
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Petroleum ether, boiling range 40 - 60 °C, reagent grade, ACS, ISO
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Petroleum Ether, Boiling Range 40–60°C, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.