Learn More
Fisherbrand™ Polystyrene Petri Dishes, Sterile
A range of high quality aseptic or sterile Petri dishes perfect for microbial or cell culture, or for sample collection, examination and storage.
Brand: Fisherbrand™ FB51510
673.92 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Description
Specifications
Polystyrene | |
Petri Dish | |
3 | |
90 mm |
Sterile | |
600 Case | |
16.2 mm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30