Learn More
Invitrogen™ eBioscience™ Permeabilization Buffer (10X)
Can be used to fix and permeabilize cells prior to performing intracellular staining of cytokines and other cytoplasmic and/or nuclear antigens.
Brand: Invitrogen™ 00-8333-56
793.28 NOK valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Alert:
Contains Sodium azide
Description
Used in conjunction with the Intracellular (IC) Fixation Buffer (cat. 00-8222) or Foxp3/Transcription Factor Fixation/Permeabilization Concentrate and Diluent (cat. 00-5521). These solutions have been specially formulated for reducing non-specific staining of fluorochrome-labeled antibodies and increasing fluorescence signal to noise ratios.
Specifications
100 mL | |
Permeabilization Reagent | |
eBioscience™ | |
Permeabilization Buffer |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2555777 | Certificate of Analysis | 21/02/2025 | 00-8333-56 |
2911220 | Certificate of Analysis | 21/02/2025 | 00-8333-56 |
2400630 | Certificate of Analysis | 21/02/2025 | 00-8333-56 |
2534041 | Certificate of Analysis | 21/02/2025 | 00-8333-56 |
2674243 | Certificate of Analysis | 21/02/2025 | 00-8333-56 |