Learn More
Perfluoro(tetradecahydrophenanthrene), mixture of isomers, Thermo Scientific Chemicals Product Code.: 11413148
Perfluoro(tetradecahydrophenanthrene), mixture of isomers, Thermo Scientific Chemicals
Product Code. 11413148
25 g, 25g
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11413148
Brand: Thermo Scientific Alfa Aesar A18898.14
Request Bulk or Custom Format
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Perfluoro(tetradecahydrophenanthrene) is used as a solvent in the preparation of co polymers obtained from emulsion polymerization of 2,3,3,3-tetrafluoroethylene and tetrafluoroethylene (TFE).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Perfluoro(tetradecahydrophenanthrene) is used as a solvent in the preparation of co polymers obtained from emulsion polymerization of 2,3,3,3-tetrafluoroethylene and tetrafluoroethylene (TFE).
Solubility
Immiscible with water.
Notes
Incompatible with lithium, sodium/sodium oxides, potassium and calcium.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 306-91-2
Molecular Formula C14F24
Molecular Weight (g/mol) 624.116
MDL Number MFCD00042596
InChI Key QKENRHXGDUPTEM-UHFFFAOYSA-N
Synonym perfluorophenanthrene, perfluoroperhydrophenanthrene, flutec pp11, perfluoro perhydrophenanthrene, tetracosafluorotetradecahydrophenanthrene, phenanthrene, tetracosafluorotetradecahydro, phenanthrene, 1,1,2,2,3,3,4,4,4a,4b,5,5,6,6,7,7,8,8,8a,9,9,10,10,10a-tetracosafluorotetradecahydro, flutec tm pp11 r, acmc-1clkp, dsstox_cid_27029
PubChem CID 78972
ChEBI CHEBI:39423
IUPAC Name 1,1,2,2,3,3,4,4,4a,4b,5,5,6,6,7,7,8,8,8a,9,9,10,10,10a-tetracosafluorophenanthrene
SMILES C12(C3(C(C(C(C1(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)(F)F)(F)F)(C(C(C(C3(F)F)(F)F)(F)F)(F)F)F)F)F
Specifications
Melting Point -20°C
Density 2.03
Boiling Point 215°C
Refractive Index 1.3348
Quantity 25 g
Solubility Information Immiscible with water.
Formula Weight 624.12
Chemical Name or Material Perfluoro(tetradecahydrophenanthrene), mixture of isomers
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number10252868 Certificate TypeCertificate of Analysis Date29/10/2024 Product CodeA18898.14,A18898.06,A18898.22
Lot Number10251000 Certificate TypeCertificate of Analysis Date30/08/2024 Product CodeA18898.14,A18898.06,A18898.22
Lot Number10196255 Certificate TypeCertificate of Analysis Date30/08/2024 Product CodeA18898.14,A18898.06,A18898.22
Lot Number10240126 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeA18898.14,A18898.06,A18898.22
Lot Number10168671 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeA18898.14,A18898.06,A18898.22
Product Title
Perfluoro(tetradecahydrophenanthrene), mixture of isomers, Thermo Scientific Chemicals >