missing translation for 'onlineSavingsMsg'
Learn More

Pentaphenylcyclopentadiene, 99%, Thermo Scientific Chemicals

Product Code. 11342219
Click to view available options
Quantity:
250 mg
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene (PPCP) was used as dopant for poly(3-alkylthiophene) diodes to enhance their electroluminescence efficiency, in the preparation of monodisperse and highly fluorescent silica nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene (PPCP) was used as dopant for poly(3-alkylthiophene) diodes to enhance their electroluminescence efficiency, in the preparation of monodisperse and highly fluorescent silica nanoparticles.

Solubility
Soluble in pyridine 5 mg/ml.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2519-10-0
Molecular Formula C35H26
Molecular Weight (g/mol) 446.593
MDL Number MFCD09909572
InChI Key YGLVWOUNCXBPJF-UHFFFAOYSA-N
PubChem CID 635023
IUPAC Name (2,3,4,5-tetraphenylcyclopenta-1,4-dien-1-yl)benzene
SMILES C1=CC=C(C=C1)C2C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Melting Point 254°C to 257°C
Quantity 5 g
Synonym 1,2,3,4,5-pentaphenyl-1,3-cyclopentadiene, pentaphenylcyclopentadiene, cyclopenta-1,3-diene-1,2,3,4,5-pentaylpentabenzene, 2,3,4,5-tetraphenylcyclopenta-1,4-dien-1-yl benzene, benzene,1,1',1,1',1-1,3-cyclopentadiene-1,2,3,4,5-pentayl pentakis, 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-yl benzene, acmc-1cek1, 2,3,4,5-tetraphenyl-1-cyclopenta-1,4-dienyl benzene, benzene, 1,1',1,1',1-1,3-cyclopentadiene-1,2,3,4,5-pentayl pentakis, 2,3,4,5-tetraphenylcyclopenta-2,4-dienyl benzene
Solubility Information Soluble in pyridine 5mg/ml.
Formula Weight 446.59
Percent Purity 99%
Chemical Name or Material Pentaphenylcyclopentadiene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'