Learn More
3M™ PELTOR™ Professional In Ear Communication Headset PIC-100 EU
The 3M™ PELTOR™ Professional In-Ear Communication Headset, PIC-100 is a next generation protective communication solution. The innovative, on-demand fit test informs the users of their personal protection level, while an in-ear speech microphone and multiple wireless technologies enable clear communication in noisy environments.
Brand: 3M™ 7100262855
9589.64 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- In-ear protective communication solution for hands-free, clear communication in noisy environments
- On-demand fit test provides the wearer's current personal attenuation rating (PAR) to help ensure adequate protection with each fit
- Speech microphones in the earpieces pick up your voice, eliminating the need for a traditional boom microphone
- The in-ear speech-communication microphone enables compatibility with a variety of personal protective equipment such as helmets/hard hats, eye wear, and respirators
- Bluetooth™ technology for connection to external devices
- Close-connect wireless technology enables close-proximity, headset-to-headset communication
- Six separate Close-connect channels enable different groups to communicate in close proximity without interference
- Environmental listening technology assists workers to hear environmental sounds in low noise and helps provide protection when it's loud
- Replaceable eartips in multiple types and sizes
- The PIC-100 introduces multiple innovative technologies to help workers stay protected and productive.
Specifications
Headset | |
None | |
0/1 V |
None | |
20/Cs. |