Specifications
Color | Black |
For Use With (Application) | Suitable Choice for Environments With Substantial Industrial Noise or Construction Machinery, Like Airports and Agricultural Work |
Plug Shape | Oval |
Product Type | Helmet Mounting Optime II |
Style | Helmet-mounted |
Includes | Helmet Mounted Ear Headphones |
Product Content Correction
Product Title
3M™ PELTOR™ Optime™ II Helmet Mounted Ear Muffs >