Learn More
3M™ PELTOR™ Optime™ I Earmuffs
PELTOR™ Optime™ I Ear Muffs are very lightweight, providing high wearer comfort. Ideal for protection against noise arising from a wide range of applications in the workplace and leisure activities. Offer versatile protection for both short and long duration tasks.
Brand:  3M™ 7000038206
2248.91 NOK valid until 2025-06-08
Use promo code "24131" to get your promotional price.
 View more versions of this product
Product Code. 15709697
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Low-profile design which minimizes obtrusiveness
  • Large space inside cup helps reduce moisture and heat build-up
  • Easy-to-replace cushions and inserts helps keep them hygienically clean
  • Soft, wide cushions help reduce pressure around the ears and improve comfort and wearability
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hi-Viz
H510P3E-469-GB
Ear Muffs
CE Approved, EN 352-3: 2002
26 dB
Hard Hat
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
Product Content Correction
Product Title
3M™ PELTOR™ Optime™ I Earmuffs > Ear Muffs; Model: H510P3E-469-GB; Style: Helmet-mounted