Learn More
3M™ PELTOR™ Hygiene Kit
3M™ PELTOR™ Hygiene Kit contains replacement sealing rings and muffler pads which can be replaced in a few easy steps.
53.64 NOK - 281.00 NOK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
15739967
|
3M™
7000108176 |
Bull's Eye™ I Earmuffs | N/A | |||||||
15749967
|
3M™
7000108177 |
Bull's Eye™ II Earmuffs | N/A | |||||||
15719957
|
3M™
7000108017 |
Bull's Eye III Earmuffs | N/A | |||||||
15729957
|
3M™
7000108018 |
H6 Series Earmuffs | N/A | |||||||
15779707
|
3M™
7000039638 |
Optime™ I Earmuffs | N/A | |||||||
15709647
|
3M™
7000009686 |
Optime™ II Earmuffs | N/A | |||||||
15709957
|
3M™
7000108016 |
Optime™ III Earmuffs |
166.00 NOK
Each |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||||
15719927
|
3M™
7000107779 |
Optime Push To Listen Earmuffs | N/A | |||||||
15710108
|
3M™
7100101874 |
Worktunes Headset |
281.00 NOK
Each |
Estimated Shipment: 11-08-2025
Log in to see stock. |
||||||
Description
- Kit contains replacement sealing rings and muffler pads which can be replaced in a few easy steps
- Extends the life of earmuffs
- Ensures good function and hygiene
- Avoid wearing uncomfortable earmuffs
- Pack contains 2 x foam inserts and 2 sealing rings
Specifications
Hygiene Kit | |
Foam Inserts and Sealing Rings |
Product Title
3M™ PELTOR™ Hygiene Kit