Learn More
3M™ PELTOR™ Cable with Stereo Plug
3M™ PELTOR™ Cable with Stereo Plug is suitable for use with 3M™ PELTOR™ Alert FM Radio and XP Headsets.
Brand:  3M™ 7000108359
203.64 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15769987
  • 287.00 NOK / Each
Only null left
Estimated Shipment: 12-08-2025
Description
  • 3.5 mm stereo connection cable
  • Includes a stereo plug
  • For use with 3M™ PELTOR™ Alert FM Radio and XP Headsets
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cable
Stereo 3.5 mm Plug
Certificates
Promotions
Promotions
Product Content Correction
Product Title
3M™ PELTOR™ Cable with Stereo Plug > Type: Cable