Learn More
Cytiva PEEK Union
Connect piping of varying sizes. Cytiva Life Sciences™ PEEK Unions are an important accessory for various scientific applications.
Brand: Cytiva 28954326
Description
- Packsize 5
- When ordered separately the connectors are black.
- Made of polyetheretherketone (PEEK)
Specifications
Union | |
1/16 Male | |
PEEK | |
0.5 mm |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%