man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Chromatography Columns and Cartridges
Ion Chromatography Columns
Anion-Exchange Packed IC Columns
Hamilton™ PEEK RCX-30
10186870
Print
Hamilton™ PEEK RCX-30
Column capacity: 1.2meq/column
Brand:
Hamilton™ 79370
View more versions of this product
Product Code.
10186870
12485.00 NOK
/ Each
Quantity
Estimated Shipment: 11-11-2024
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Specifications
Column Capacity
1.2 meq/column
Particle Size
7 μm
Stationary Phase
RCX-30