missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD 158780, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent inhibitor of ErbB receptor family tyrosine kinases (IC50 values are 0.008, 49 and 52 nM for EGFR, ErbB2 and ErbB2/ErbB4 respectively) that does not inhibit FGF or PDGF-mediated tyrosine phosphorylation. Induces G1 cell cycle arrest in MCF10A cells and is antiproliferative in A431 human epidermal carcinoma cells.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 330.18 |
Percent Purity | >99% |
Chemical Name or Material | PD 158780 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'