Learn More
BRAND™ 8-Strip PCR Caps, BIO-CERT™ PCR QUALITY
BRAND™ PCR cap strips ensure reliable closures for PCR strips and plates, safeguarding your samples against evaporation and contamination.
Brand: BRAND™ 781340
304.18 NOK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
Description
- Exceptional Quality and Purity: BIO-CERT™ PCR QUALITY ensures caps are free from human DNA, RNase, DNase, pyrogens, and PCR inhibitors.
- Secure Sealing: Designed to reliably seal PCR strips and plates with 24-, 48-, or 96-wells, effectively protecting your samples from evaporation and contamination.
- Perfect Compatibility: Optimally shaped to match BRAND™ PCR tube strips and plates, ensuring easy opening and closing. They are also compatible with most other manufacturers, offering flexibility for your workflow.
- Contamination Prevention: Equipped with side tabs for precise positioning and a small opening attachment on each cap to prevent direct contact with the interior.
- qPCR-Ready Transparency: Highly transparent flat caps enable accurate fluorescence measurement, making them ideal for qPCR applications.
- Enhanced Sealing with Domed Caps: Domed caps provide a stronger sealing effect when used with thermal cyclers lacking pressure sensors.
Specifications
Clear | |
Domed | |
Polypropylene | |
Yes | |
Autoclavable | |
Single-use |
Thermal cycler | |
Non-sterile | |
Yes | |
8-strip Domed Cap | |
Yes | |
Cap Strip |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
0000013669 | Certificate of Analysis | 06/06/2025 | 781340 |
0000012878 | Certificate of Analysis | 25/11/2024 | 781340 |
0000012400 | Certificate of Analysis | 18/10/2024 | 781340 |
Promotions
-
Multiple Manufacturers
Genomics Powerhouse
Power your pipeline from PCR to sequencing.Promo Code: 24117Expires: 2025-08-24
New Offer Save Up to 92%