missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCID2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | PCID2 |
---|---|
Applications | Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17501356
|
Proteintech
26638-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17591346
|
Proteintech
26638-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PCID2 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human PCID2. This antibody recognizes human antigen. The PCID2 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
PCID2 | |
Polyclonal | |
Rabbit | |
55795 | |
Primary |
Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title