Learn More
Patron Photometer Kit, Palintest™ Product Code.: 11993707
Patron Photometer Kit, Palintest™
Product Code. 11993707
Photometer Kit
Click to view available options
:
Photometer Kit
Product Code. 11993707
Brand: Palintest PT804
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Type Check Standard
compliance-icons compliance-icons
Product Identifier
  • Check Standards Palintest with test certificate
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1
  • Respiratory sensitiser Category 1
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 1A
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Reproductive toxicity Category 1B
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H350-May cause cancer.
  • H317-May cause an allergic skin reaction.
  • H360F-May damage fertility.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H360D-May damage the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH208-Contains potassium hexachloroplatinate. May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ first aider.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P391-Collect spillage.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acidcobalt dichloride
Product Content Correction
Product Title
Patron Photometer Kit, Palintest™ >