Learn More
Amarell™ Partial Immersion Eco-friendly Thermometer
Brand: Amarell™ G11388
This item is not returnable.
View return policy
Specifications
Green Liquid | |
Partial Immersion | |
Partial Immersion Thermometer | |
-10°C | |
1°C |
General use | |
300 mm | |
110°C | |
-10°C to +110°C | |
76 mm |
Product Content Correction
Product Title
Amarell™ Partial Immersion Eco-friendly Thermometer > Temp. Range: -10 to 110deg.C; Subdivision: 1deg.C