Learn More
Cole-Parmer™ Parker Hannifin™ Metering Valve with Compression Connections
No need to worry about repeating flowrates with these valves
Brand:  Cole-Parmer™ 98450-25
Product Code. 11780429
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • These valves feature an analog knob for repeatable flow settings.
  • Brass valves have Buna N O-rings.
  • 316 SS valves have Viton O-rings.
Specifications
Specifications
16 SS Metering Valve with compression connections, 1/4in tube OD, 0.055 in. orifice
Stainless Steel
0.25 in. OD
1/4 in. Compression
204°C
Compression Connection
0.25 in.
Air, Max: 450L/min, Water, Max 3.2L/min
400°F
Documents
One moment while we fetch your results.
Product Title
Cole-Parmer™ Parker Hannifin™ Metering Valve with Compression Connections > Tubing size: 0.25 in. OD