Learn More
MP Biomedicals Pantothenic Acid Deficient Diet, Modified, Pellet
Pantothenic acid deficient diet for rabbits. Lacks pantothenic acid.
Brand:  MP Biomedicals 0296015910
 View more versions of this product
Product Code. 11472851
  • 9105.00 NOK / 10kg
Explore more special offers
Description
Description
  • Pantothenic acid control diet for rabbits
  • Lacks pantothenic acid
  • Contains casein, methionine, mineral mix and MP Bio vitamin fortification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Single-use
10 kg
Room Temperature
0.3048 m
Animal Diet
Pellet
Non-sterile
1 ft.
Special Offers
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.