missing translation for 'onlineSavingsMsg'
Learn More

Palmitelaidic acid, >99%, MP Biomedicals™

Product Code. 11429361
Click to view available options
Quantity:
100 mg
0.5 g
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 61-25-6
Molecular Formula C20H22ClNO4
Molecular Weight (g/mol) 375.849
InChI Key UOTMYNBWXDUBNX-UHFFFAOYSA-N
Synonym papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan
PubChem CID 6084
IUPAC Name 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride
SMILES COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl

Specifications

Color Colorless
Density 0.895g/mL at 20°C
Assay Percent Range ≥99%
Quantity 100 mg
Formula Weight 254.4
Percent Purity >99%
Physical Form Frozen Liquid
Chemical Name or Material Palmitoleic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.