Learn More
PAL International™ PAL TX Surface Disinfectant Wipes Pocket Probe Canister
A small pocket size portable dispensing canister for convenient disinfection of probes and utensils
Brand: PAL International™ W65231T
This item is not returnable.
View return policy
Description
- Perfect for laboratory testing in food factories – both at production line side and in the lab itself
- Can also be easily stored for use in food retail outlets where temp probes are used regularly
- Pal TX Surface Disinfectant wipes are food safe and proven to kill most microorganisms such as bacteria, yeast & viruses
- Pal TX Surface Disinfectant Wipes clean and disinfect without using Quats, PHMB or alcohol
Specifications
Pocket Canister | |
70 x 130 mm | |
Disinfection of probes and utensils | |
70 Wipes/Canister, 48 Canisters/Pk. |