Learn More
Tocris Bioscience™ PACMA 31
Irreversible protein disulfide isomerase (PDI) inhibitor
Brand:  Tocris Bioscience™ 5116/10
1058.29 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18787048
  • 1200.00 NOK / 10mg
Explore more special offers
Description
Description
PACMA 31 is an irreversible protein disulfide isomerase (PDI) inhibitor; inhibits growth of ovarian cancer cells. Suppresses tumor growth in a mouse xenograph model of human ovarian cancer.
Specifications
Specifications
>99%
PACMA 31
PACMA 31
10 mg
430.47
Special Offers
For Research Use Only