Learn More
Buerkle™ Packaging Buckets
Manufactured from polypropylene for strength and durability. Buerkle™ Packaging Buckets are ideal for transport, packaging and storing of a wide range of materials.
23.40 NOK - 78.60 NOK
Specifications
Product Type | Packaging Bucket |
---|---|
Material | Polypropylene |
Color | White |
For Use With (Equipment) | Suitable for The Food or Building Material Industries and Non-Aggressive Chemicals in Environmental Protection, Hospitals and The Household |
Includes | Quality Closure and Strong Lid |
Product Code | Brand | Capacity (Metric) | Diameter (Metric)Top | Height (Metric) With Closure | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric)Top | Height (Metric) With Closure | Price | Quantity & Availability | ||||
11831091
|
Buerkle™
2327-0001 |
1 L | 132 mm | 131 mm |
23.40 NOK
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||
11841091
|
Buerkle™
2327-0020 |
2 L | 175 mm | 132 mm |
36.55 NOK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||
11851091
|
Buerkle™
2327-0030 |
3 L | 225 mm | 123 mm |
46.10 NOK
Each |
In Stock
Log in to see stock. ![]() |
||||
11861091
|
Buerkle™
2327-0050 |
5 L | 225 mm | 194 mm |
58.40 NOK
Each |
In Stock
Log in to see stock. ![]() |
||||
11871101
|
Buerkle™
2327-0010 |
10 L | 268 mm | 263 mm |
78.60 NOK
Each |
In Stock
Log in to see stock. ![]() |
||||
Description
Suitable for food or building material industries and non-aggressive chemicals in environmental protection, hosptials and household. Features strong handle and quality closure for product safety.- White polypropylene
- Air-tight closure
- Includes lid
Specifications
Packaging Bucket | |
White | |
Quality Closure and Strong Lid |
Polypropylene | |
Suitable for The Food or Building Material Industries and Non-Aggressive Chemicals in Environmental Protection, Hospitals and The Household | |
1 Ea. |
Product Title
Buerkle™ Packaging Buckets