missing translation for 'onlineSavingsMsg'
Learn More

VACUUBRAND™ Package Rotary Vane Pump RZ 6 with Vacuum Gauge VACUU·VIEW Extended
GREENER_CHOICE

The rotary vane pump package RZ 6 with oil mist filter, butterfly valve VS16 and vacuum gauge VACUU·VIEW extended offers great performance in a compact design - even with the gas ballast open, they enable a very good ultimate vacuum.

52605.00 NOK

Specifications

Max. Vacuum 0.002 mbar
Flow Rate 5.7 m3/hr.
Pump Type Rotary Vane Vacuum Pump
Enclosure Rating IP40
Applications Degassing, Lyophilization, Fine Vacuum Distillation and Drying Chambers
View More Specs

Products 1
Product Code Brand Plug Type Price Quantity & Availability  
Product Code Brand Plug Type Price Quantity & Availability  
17851157
GREENER_CHOICE
View Documents
VACUUBRAND™
20798160
CEE
52605.00 NOK
Each
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
Description

Description

  • High flow rates even at vacuum levels approaching ultimate vacuum
  • High water vapor tolerance due to efficient gas ballast; very good ultimate vacuum even with gas ballast
  • Vacuum-tight at switch-off; external anti-suckback valve not needed
  • Large oil volume: Long intervals between oil changes
  • Ease of maintenance due to telescopic design
Specifications

Specifications

0.002 mbar
Rotary Vane Vacuum Pump
Degassing, Lyophilization, Fine Vacuum Distillation and Drying Chambers
230 V
5.7 m3/hr.
IP40
1.1 bar
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.