missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2RY13 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20335-1-AP-150UL
This item is not returnable.
View return policy
Description
The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is activated by ADP.Specifications
P2RY13 | |
Polyclonal | |
Unconjugated | |
P2RY13 | |
FKSG77, GPCR1, GPR86, GPR94, P2RY13, P2Y13, SP174 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
53829, 74191 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BPV8, Q9D8I2 | |
P2RY13 | |
P2RY13 Fusion Protein Ag14108 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction