Learn More
HALYARD™ PUREZERO™ Pouch Face Mask with Headband Product Code.: 17857980
Only undefined left
Add to basket
HALYARD™ PUREZERO™ Pouch Face Mask with Headband
GREENER_CHOICE
Product Code. 17857980 Shop All Halyard Products
with Headband
Click to view available options
:
with Headband
Product Code. 17857980
Brand: HALYARD™ CLN3941BBG
Only undefined left
Add to basket
Only undefined left
Add to basket
A three-layer mask that provides high particle, bacterial, viral filtration efficiency, and expanded breathing chamber. The mask consists of SO SOFT biocomponent technology inner lining for comfort and low linting neckguard for increased protection.
  • Designed to meet the stringent requirements of cleanroom environments
  • Manage user comfort and protection with SO SOFT inner lining, a foam nose band for comfort and fog reduction
  • Manage product contamination by providing high particle, bacterial, and viral filtration efficiency
Greener Choice Claims
  • Environmental benefits include:
    • Recycled Content
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Headband
Material Polyester, Polyethylene
Size One Size
Sterility Non-sterile
Style Pouch
Certifications/Compliance General Product Safety Directive 2001/95/EC, ISO5/6/7/8
Content And Storage Store in a clean, dry environment. Avoid direct sunlight or other sources of heat.
Disposable Single-use
Filter Efficiency BFE > 98%, PFE > 98%,VFE 97%
For Use With (Application) Cleanrooms
Latex Free Yes
Model CLN3941
Packaging Quantity 50/Bag, 10 Bags/Cs.
Packaging Type PE Bag
Width (English) 10 in.
Product Title
HALYARD™ PUREZERO™ Pouch Face Mask with Headband >