missing translation for 'onlineSavingsMsg'
Learn More
p-Toluenesulfonamide, 99% Product Code.: 10104293

p-Toluenesulfonamide, 99%

Product Code. 10104293 Shop All Thermo Scientific Chemicals Products
5 kg, Plastic drum, 5kg
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 10104293

Brand: Thermo Scientific Acros 147690050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70-55-3 | C7H9NO2S | 171.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-55-3
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00011692
InChI Key LMYRWZFENFIFIT-UHFFFAOYSA-N
Synonym p-toluenesulfonamide, 4-toluenesulfonamide, tosylamide, p-tosylamide, benzenesulfonamide, 4-methyl, p-tolylsulfonamide, toluene-4-sulfonamide, tolylsulfonamide, 4-methylbenzene-1-sulfonamide, p-toluenesulfamide
PubChem CID 6269
ChEBI CHEBI:34435
IUPAC Name 4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N

Specifications

Melting Point 136.0°C to 140.0°C
Color White
Boiling Point 221.0°C (10.0 mmHg)
Flash Point 202°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic drum
Linear Formula CH3C6H4SO2NH2
Quantity 5 kg
Beilstein 11, 104
Fieser 13,294
Solubility Information Solubility in water: 0.32 g/100 mL(25° c). Other solubilities: soluble in alcohol
Formula Weight 171.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material p-Toluenesulfonamide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Toluenesulfonamide, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.