missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Nitrophenethyl alcohol, 98.1%, MP Biomedicals™
Click to view available options
Quantity:
10 g
Specifications
CAS | 100-27-6 |
Color | Yellow |
Assay Percent Range | 0.981 |
Molecular Formula | C8H9NO3 |
Quantity | 10 g |
Synonym | 4-nitrophenethyl alcohol, 2-4-nitrophenyl ethanol, 4-nitrobenzeneethanol, 2-p-nitrophenyl ethanol, benzeneethanol, 4-nitro, p-nitrophenethyl alcohol, 2-4-nitrophenyl ethan-1-ol, phenethyl alcohol, p-nitro, ccris 6079, 4-nitrophenethylalcohol |
InChI Key | IKMXRUOZUUKSON-UHFFFAOYSA-N |
SMILES | C1=CC(=CC=C1CCO)[N+](=O)[O-] |
IUPAC Name | 2-(4-nitrophenyl)ethanol |
Molecular Weight (g/mol) | 167.164 |
Afficher plus |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'