missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxytetracycline Dihydrate 95%

1245.00 NOK - 8330.00 NOK

Chemical Identifiers

CAS 6153-64-6
Molecular Formula C22H28N2O11
Molecular Weight (g/mol) 496.469
MDL Number MFCD00151234
InChI Key IMLJLCJZQLGHJS-JEKSYDDFSA-N
PubChem CID 54680691
IUPAC Name (4S,4aR,5S,5aR,6S,12aR)-4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;dihydrate
SMILES CC1(C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O)O.O.O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10631925
SDS View Documents
Thermo Scientific Chemicals
123840100
10 g Glass Bottle
1245.00 NOK
10g
Please to purchase this item. Need a web account? Register with us today!
10356910
SDS View Documents
Thermo Scientific Chemicals
123841000
100 g Glass Bottle
8330.00 NOK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Oxytetracycline Dihydrate 95%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.