Learn More
Thermo Scientific™ Oxoid™ X & V Factor Discs
For identification of Haemophilus spp.
Brand: Thermo Scientific™ DD0005B
640.41 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
For identification of Haemophilus spp.
50 Disks/Cartridge
Specifications
Haemophilus species | |
V Factor Disc | |
5 x 50 Discs |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6215241 | Certificate of Analysis | 21/02/2025 | DD0005B |
6199997 | Certificate of Analysis | 19/12/2024 | DD0005B |
6130673 | Certificate of Analysis | 18/07/2024 | DD0005B |
6130674 | Certificate of Analysis | 18/07/2024 | DD0005B |
3704935 | Certificate of Analysis | 06/01/2024 | DD0005B |