Learn More
Thermo Scientific™ Oxoid™ X Factor Discs
Perform identification of Haemophilus spp. with Thermo Scientific™ Oxoid X Factor Discs.
Brand: Thermo Scientific™ DD0003B
627.55 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Perform identification of Haemophilus spp. with Thermo Scientific™ Oxoid X Factor Discs.
Ordering Alerts:
50 Discs/Cartridge
Specifications
-10°C to -20°C | |
Haemophilus species | |
5 Cart./Pk. |
X Factor Discs | |
X Factor Disc | |
50 Disks/Cartridge |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6224258 | Certificate of Analysis | 21/02/2025 | DD0003B |
6188874 | Certificate of Analysis | 13/12/2024 | DD0003B |
6130672 | Certificate of Analysis | 19/07/2024 | DD0003B |
6110072 | Certificate of Analysis | 19/06/2024 | DD0003B |
3803876 | Certificate of Analysis | 09/05/2024 | DD0003B |