Learn More
Thermo Scientific™ Oxoid™ Sheep Blood
Brand:  Thermo Scientific™ SR0051B
163.17 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12967755
  • 221.00 NOK / 25mL
Specifications
25 mL
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Sheep Blood > Quantity: 25mL