Learn More
Thermo Scientific™ Oxoid™ Penicillin G Antimicrobial Susceptibility discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Brand:  Thermo Scientific™ CT0152B
305.27 NOK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
 View more versions of this product
Product Code. 12761760
  • 333.00 NOK / Pack of 250
Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Penicillin G Antimicrobial Susceptibility discs.
Ease of use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results, but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Penicillin G
1 unit
P
Antimicrobial Susceptibility
50 Discs/Cartridge
Complies with BSAC, EUCAST guidelines
Oxoid Penicillin G Antimicrobial Susceptibility Discs
Antimicrobial Susceptibility Discs
5 Cartridges/Pk.
50 Discs/Cartridge
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6259592 Certificate TypeCertificate of Analysis Date08/05/2025 Catalog NumberCT0152B
Lot Number6244950 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog NumberCT0152B
Lot Number6230188 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog NumberCT0152B
Lot Number6220796 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberCT0152B
Lot Number6220794 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberCT0152B
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Penicillin G Antimicrobial Susceptibility discs >