Learn More
Thermo Scientific™ Oxoid™ IMAGEN™ Chlamydia Positive Control Slide
Oxoid IMAGEN Chlamydia Positive Control Slide contains acetone-fixed human epithelial cells infected with Chlamydia.
Brand: Thermo Scientific™ S610930-2
900.68 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The IMAGEN™ Chlamydia Control Slides are for use as a control for the IMAGEN™ Chlamydia test (Code No. K610111-2).
Sealed in foil pouch filled with nitrogen.
Specifications
IMAGEN™ Chlamydia Positive Control Slide | |
Chlamydia | |
Chlamydia Control Slide | |
1 Slide/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6245573 | Certificate of Analysis | 14/06/2025 | |
6135731 | Certificate of Analysis | 08/11/2024 | S610930-2 |
6127317 | Certificate of Analysis | 30/05/2024 | S610930-2 |
3770894 | Certificate of Analysis | 13/12/2023 | S610930-2 |
3726323 | Certificate of Analysis | 13/10/2023 | S610930-2 |