Learn More
Thermo Scientific™ Oxoid™ Gentamicin Antimicrobial Susceptibility Discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Brand: Thermo Scientific™ CT0794B
267.80 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Gentamycin Antimicrobial Susceptibility discs.
- Easy-to-read labeling
- Shatterproof, spring-loaded cartridges
- End-of-cartridge indicator
- Reproducible performance
Ordering Alerts:
50 Discs/Cartridge; 5 Cartridges/Package. Sold by 5 Cartridges/Package only.
Specifications
Gentamicin | |
120µg | |
CN | |
Antimicrobial Susceptibility | |
50 Discs/Cartridge |
Complies with CLSI guidelines | |
Oxoid Gentamicin Antimicrobial Susceptibility Discs | |
Antimicrobial Susceptibility Discs | |
5 Cartridges/Pk. | |
50 Discs/Cartridge |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6242212 | Certificate of Analysis | 21/03/2025 | CT0794B |
6191426 | Certificate of Analysis | 20/11/2024 | CT0794B |
6178072 | Certificate of Analysis | 18/10/2024 | CT0794B |
6121983 | Certificate of Analysis | 14/06/2024 | CT0794B |
3809800 | Certificate of Analysis | 22/03/2024 | CT0794B |