Learn More
Thermo Scientific™ Oxoid™ Fraser Supplement
Oxoid Fraser Supplement is used for the isolation of Listeria species.
Brand: Thermo Scientific™ SR0156E
1292.76 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Oxoid Fraser Supplement is used for the isolation of Listeria species.
- Add to Fraser Broth Base, Part No. CM0895B
- Each vial supplements 500mL of medium
Specifications
Fraser Supplement | |
Microbiology Media | |
Each vial for 500mL medium |
Freeze-dried | |
10/Pk. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
16Hours
8Minutes
47Seconds
46 -
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
Days
57Hours
8Minutes
47Seconds
46