Learn More
Thermo Scientific™ Oxoid™ AnaeroJar™ Base, Jar
Oxoid AnaeroJar Base is a polycarbonate jar base for use with AnaeroJar 2.5L, Part No. AG0025A.
Brand: Thermo Scientific™ AG0026A
1961.50 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Oxoid AnaeroJar Base is a polycarbonate jar base for use with AnaeroJar 2.5L, Part No. AG0025A.
Specifications
AnaeroJar Base | |
Oval Jar | |
Thermo Scientific Oxoid | |
Each |
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ AnaeroJar™ Base, Jar > Jar, Base