Learn More
Oxalic Acid, 0.05M, Reagecon™ Product Code.: 11556813
Oxalic Acid, 0.05M, Reagecon™
BIOPHARMA
Product Code. 11556813 Shop All Reagecon Products
1 L, 1 L
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. 11556813
Brand: Reagecon OA20051
Specifications
Quantity 1 L
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Product Code
Lot NumberOA2525E1 Certificate TypeCertificate of Analysis Date11/06/2025 Product CodeOA20051
Lot NumberOA2524F1 Certificate TypeCertificate of Analysis Date10/07/2024 Product CodeOA20051
Lot NumberOA2522C1 Certificate TypeCertificate of Analysis Date05/08/2022 Product CodeOA20051
Lot NumberOA2520M1 Certificate TypeCertificate of Analysis Date01/03/2022 Product CodeOA20051
Product Content Correction
Product Title
Oxalic Acid, 0.05M, Reagecon™ >