missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid, 0.005 M (0.01 N), Scharlab

Product Code. 11368624
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 144-62-7
Molecular Formula C2H2O4
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
SMILES OC(=O)C(O)=O
IUPAC Name oxalic acid
Molecular Weight (g/mol) 90.03
Quantity 1 L
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'