Learn More
Thermo Scientific Chemicals Oxacillin sodium salt, 95%
Oxacillin sodium salt, CAS # 1173-88-2, is an antibiotic that is used primarily in studying gram-positive bacteria.
614.00 NOK - 1820.00 NOK
Chemical Identifiers
CAS | 1173-88-2 |
---|---|
Molecular Formula | C19H21N3NaO5S |
Molecular Weight (g/mol) | 426.443 |
InChI Key | IFFSBWVMGGQKTD-VICXVTCVSA-N |
PubChem CID | 131673929 |
IUPAC Name | (2S,5R,6R)-3,3-dimethyl-6-[(5-methyl-3-phenyl-1,2-oxazole-4-carbonyl)amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium |
SMILES | [HH].CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.[Na] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15345008
|
Thermo Scientific Chemicals
455440010 |
1 g |
614.00 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15355008
|
Thermo Scientific Chemicals
455440050 |
5 g |
1820.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Oxacillin sodium salt is a narrow-spectrum beta-lactam antibiotic
- It inhibits biosynthesis of bacterial cell walls at the level of transpeptidation (PBP) of peptidoglycans
Applications
- Oxacillin sodium salt is an antibiotic that is active against gram-positive bacteria
- Oxacillin can be used in studies of mechanisms of resistance to penicillinase and antimicrobial susceptibility
- It has also been used to study autolysins
Chemical Identifiers
1173-88-2 | |
426.443 | |
131673929 | |
[HH].CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.[Na] |
C19H21N3NaO5S | |
IFFSBWVMGGQKTD-VICXVTCVSA-N | |
(2S,5R,6R)-3,3-dimethyl-6-[(5-methyl-3-phenyl-1,2-oxazole-4-carbonyl)amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium |
Specifications
Oxacillin Sodium Salt | |
Authentic | |
C19H21N3NaO5S | |
IFFSBWVMGGQKTD-VICXVTCVSA-N | |
(2S,5R,6R)-3,3-dimethyl-6-[(5-methyl-3-phenyl-1,2-oxazole-4-carbonyl)amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium | |
131673929 | |
95% |
1173-88-2 | |
3.5 to 5.0% | |
Solubility in water: 50g/L. | |
[HH].CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.[Na] | |
426.443 | |
423.42 | |
Oxacillin sodium salt |
Safety and Handling
- Oxacillin sodium salt
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.