Learn More
Thermo Scientific™ Owl™ EasyCast™ B2 and B3 System Replacement Parts
Replace your UVT gel tray, gaskets and external casters with B2 and B3 system replacement parts.
211.00 NOK - 5515.00 NOK
Specifications
For Use With (Equipment) | Owl™ EasyCast B2 and B3 Mini Gel Electrophoresis System |
---|---|
Gel Size | Mini |
Product Line | Owl, EasyCast |
Product Code | Brand | Type | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | Price | Quantity & Availability | |||||
11932965
|
Thermo Scientific™
B2-CST |
External Caster | Trays and Combs Not Included | N/A | ||||||
11942965
|
Thermo Scientific™
B2-GK |
Gasket | Gasket Pair |
211.00 NOK
2 sets |
Estimated Shipment: 14-08-2025
Log in to see stock. |
|||||
11947585
|
Thermo Scientific™
B2-RL |
Gel Tray | 12 Slots, 12 Rapid Load Combs |
5515.00 NOK
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
11972965
|
Thermo Scientific™
B2-UVT |
Gel Tray | Gel Tray Only |
1230.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
Description
Replace your UVT gel tray, gaskets and external casters with Thermo Scientific™ Owl™ EasyCast™ B2 and B3 System Replacement Parts, for use with EasyCast™ Mini Gel Systems.
For use with Owl EasyCast B2 and B3 Gel Systems.Specifications
Owl™ EasyCast B2 and B3 Mini Gel Electrophoresis System | |
Mini | |
Owl, EasyCast |