Learn More
Uvex™ Oversleeves 9862 PE White 50 pair
Gauntlets, Disposable chemical protection. Material: PP spunbond laminated with PE film
Brand: Uvex™ 9875300
500.63 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Gauntlets Disposable chemical protection Accessories
- Color: white
- Material: PP spunbond laminated with PE film
- Elastic on both sides
- 50 pairs in polybag
- For single use only
- Size: one size fits all
Specifications
Universal | |
Non-woven Microporous Polyethylene Laminate | |
Non-sterile | |
Reusable | |
60 g/m² |
Chemical-resistant Sleeve | |
White | |
Standard Sleeve | |
Taped |
Product Content Correction
Product Title
Uvex™ Oversleeves 9862 PE White 50 pair > Size: One size